Order Entry
Italy
ContactUsLinkComponent
HEATING MANTLES WHM, FOR 2000 ML REACTION VESSELS, CYLINDRICAL, A-Ø 130 MM, HEIGHT 190 MM, INSIDE DEPTH: 140 MM, WITHOUT POWER CONTROLLER, TEMP. RANGE 450°C, 450 W, ALUMINIUM CASING, POWER SUPPLY 230 V, SERIAL NO. 1 * 1 items
Catalog # WITGDH.WHM12213
Supplier:  witeg Labortechnik
HEATING MANTLES WHM, FOR 2000 ML REACTION VESSELS, CYLINDRICAL, A-Ø 130 MM, HEIGHT 190 MM, INSIDE DEPTH: 140 MM, WITHOUT POWER CONTROLLER, TEMP. RANGE 450°C, 450 W, ALUMINIUM CASING, POWER SUPPLY 230 V, SERIAL NO. 1 * 1 items
Catalog # WITGDH.WHM12213
Supplier:  witeg Labortechnik
Frequently Bought Together