Order Entry
Italy
ContactUsLinkComponent
C18P x 5µm particle size x 120Å pore size, 10mm length x 150µm ID, PEEKsil ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2POL-05G15K
C18P x 5µm particle size x 120Å pore size, 10mm length x 150µm ID, PEEKsil ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2POL-05G15K
Frequently Bought Together