Order Entry
Italy
ContactUsLinkComponent
C8H x 3µm particle size x 1000Å pore size, 10mm length x 150µm ID, PEEKsil ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C82-03G15K
C8H x 3µm particle size x 1000Å pore size, 10mm length x 150µm ID, PEEKsil ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C82-03G15K
Frequently Bought Together