Order Entry
Italy
ContactUsLinkComponent
WB SUBSTRATE KIT I WB SUBSTRATE KIT I IS A NON-ISOTOPIC, LUMINOL-BASED CHEMILUMINESCENCE SUBSTRATE, DESIGNED FOR THE CHEMILUMINESCE NT DETECTION OF IMMOBILIZED PROTEINS AND IMMOBILIZED NUCLEIC ACIDS CONJUGATED WITH HORSERADISH PEROXIDASE 1 * 1 KIT
Catalog # ABNOKA4034
Nome del fornitore:  Abnova
WB SUBSTRATE KIT I WB SUBSTRATE KIT I IS A NON-ISOTOPIC, LUMINOL-BASED CHEMILUMINESCENCE SUBSTRATE, DESIGNED FOR THE CHEMILUMINESCE NT DETECTION OF IMMOBILIZED PROTEINS AND IMMOBILIZED NUCLEIC ACIDS CONJUGATED WITH HORSERADISH PEROXIDASE 1 * 1 KIT
Catalog # ABNOKA4034
Nome del fornitore:  Abnova
Frequently Bought Together