Order Entry
Italy
ContactUsLinkComponent
POLARIMETER ADP 650 DIGITAL (UK PLUG) MULTIPLE WAVELENGTH 365,405,436,546 & 589NM 1 * 1 items
Catalog # 635-0787
Supplier:  BELLINGHAM STANLEY
undefined
POLARIMETER ADP 650 DIGITAL (UK PLUG) MULTIPLE WAVELENGTH 365,405,436,546 & 589NM 1 * 1 items
Catalog # 635-0787
Supplier:  BELLINGHAM STANLEY
Frequently Bought Together