Objectives for PSM-1000 microscopes
ELWD plan apo objective (WD = 3 mm)
Catalog # MOTI1101001700021
Supplier: MOTIC
Jump to:
Product Details & Documents
Specifications
- Magnification:100×/0,8
- Description:ELWD plan apo objective (WD = 3 mm)
- Seq:110