Objectives for PSM-1000 microscopes
ELWD plan apo objective
Catalog # MOTI1101001700112
Supplier: MOTIC
Jump to:
Product Details & Documents
Specifications
- Magnification:10×
- Description:ELWD plan apo objective
- Seq:105