IQ/OQ document with works test data for chamber, incl. Temperature distribution survey for one free-selectable temperature and humid ity value. 27 measuring points to DIN 12 880: 2007-05, PQ check list as support for validation by customer (models HP 1 * 1 items
Catalog # MEMMD00136ICH260
Nome del fornitore: MEMMERT
Frequently Bought Together