Order Entry
Italy
ContactUsLinkComponent
Dialysis tubing Trial kit Spectra/Por 7 pre-treated standard regenerated cellulose (RC) membrane MWCO 50kD 28mm flat width (18mm dia meter) 2,5ml/cm including tubing closures and openers wet 1 * 1 m
Catalog # SPEC132129T
Dialysis tubing Trial kit Spectra/Por 7 pre-treated standard regenerated cellulose (RC) membrane MWCO 50kD 28mm flat width (18mm dia meter) 2,5ml/cm including tubing closures and openers wet 1 * 1 m
Catalog # SPEC132129T
Frequently Bought Together