Order Entry
Italy
ContactUsLinkComponent
BC8004, 800mm wide Chemcap Clearview Ductless Fume Cabinet has teardrop front working access as standard. Supplied with PVC base tray. Supplied complete with /OS Chemcap Enviro carbon filtration. Optional static/mobile bench. Supplied with FREE repla 1 * 1 items
Catalog # 135-1859
Supplier:  BIGNEAT
BC8004, 800mm wide Chemcap Clearview Ductless Fume Cabinet has teardrop front working access as standard. Supplied with PVC base tray. Supplied complete with /OS Chemcap Enviro carbon filtration. Optional static/mobile bench. Supplied with FREE repla 1 * 1 items
Catalog # 135-1859
Supplier:  BIGNEAT
Frequently Bought Together