Order Entry
Italy
Orders LinkContactUsLinkComponent
Mouse serum
Mouse serum
Mouse serum, Sterile filtered, Frozen
Catalog # S2160-010
Supplier:   Biowest
Mouse serum
Catalog # S2160-010
Supplier:  Biowest
Mouse serum
Mouse serum
Mouse serum, Sterile filtered, Frozen
Price
51,20 €
Log in for availability
1 * 10 mL
Product Details & Documents
Mouse serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Mouse serum is used as a blocking agent in immunoassays and controls.
Documents
Specifications
  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    Mouse serum
  • Seq:
    42