AdvanTip™ Pipette Tips
Low retention, 2 - 300 µl, -, 250, 300 μl pipettes, Racked, 96 tips/rack
Catalog # HAMI68009-21
Supplier: HAMILTON BONADUZ
Jump to:
Product Details & Documents
AdvanTip™ precision pipette tips are perfect for routine, high-throughput work where an extensive range of universal tips and multiple packaging options are needed to meet laboratory requirements.
- Made from chemically resistant, virgin PP for excellent clarity and smooth surfaces
- Certified free from RNase, DNase, and pyrogens
- Extended length (only some tips): Prevent sample cross-contamination and protect pipettes from contamination when pipetting into long, narrow tubes or deep well plates
- Low retention tips (only some tips): Feature walls that maximise sample recovery; ideal for quantitative PCR, valuable samples and reagents
- Universal fit, can be used with Hamilton SoftGrip™ manual pipettes
Specifications
- Packed:Racked, 96 tips/rack
- Sterile:-
- Volume:2 - 300 µl
- Seq:04
- Tip style:Low retention
- For use with:250, 300 μl pipettes