Order Entry
Italy
ContactUsLinkComponent
TFF-2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 106 amino acids (24-129) and having total molecular mass of 12 kDa. TFF2 Human Recombinant includes 40-amino acid trefoil motif containin 1 * 5 µG
Catalog # BIRBORB80105-5
Supplier:  Biorbyt
TFF-2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 106 amino acids (24-129) and having total molecular mass of 12 kDa. TFF2 Human Recombinant includes 40-amino acid trefoil motif containin 1 * 5 µG
Catalog # BIRBORB80105-5
Supplier:  Biorbyt