Order Entry
Italy
ContactUsLinkComponent
Human IL31 recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 141 amino acids (24-164 a.a.) and having a molecular mass of 15.8 kDa. 1 * 10 µG
Catalog # BIRBORB80082-10
Supplier:  Biorbyt
Human IL31 recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 141 amino acids (24-164 a.a.) and having a molecular mass of 15.8 kDa. 1 * 10 µG
Catalog # BIRBORB80082-10
Supplier:  Biorbyt