Order Entry
Italy
ContactUsLinkComponent
IL1b Rat Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 152 amino acids and having a molecular mas s of 17.3 kDa. The IL-1b is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80052-2
Supplier:  Biorbyt
IL1b Rat Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 152 amino acids and having a molecular mas s of 17.3 kDa. The IL-1b is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80052-2
Supplier:  Biorbyt