Order Entry
Italy
ContactUsLinkComponent
Interferon-alpha 1b Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 166 amino acids and having a molecular mass of 19392 Dalton.The Interferon-alpha 1b gene was obtained from human leukocytes 1 * 10 µG
Catalog # BIRBORB80037-10
Supplier:  Biorbyt
Interferon-alpha 1b Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 166 amino acids and having a molecular mass of 19392 Dalton.The Interferon-alpha 1b gene was obtained from human leukocytes 1 * 10 µG
Catalog # BIRBORB80037-10
Supplier:  Biorbyt