Order Entry
Italy
ContactUsLinkComponent
GMF-Beta (GMF-Beta) Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 141 amino acids a nd having total molecular mass of 16.5 kDa. GMF-Beta, GMF-Beta, Human Recombinant is purified by proprietary 1 * 2 µG
Catalog # BIRBORB80036-2
Supplier:  Biorbyt
GMF-Beta (GMF-Beta) Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 141 amino acids a nd having total molecular mass of 16.5 kDa. GMF-Beta, GMF-Beta, Human Recombinant is purified by proprietary 1 * 2 µG
Catalog # BIRBORB80036-2
Supplier:  Biorbyt