Order Entry
Italy
ContactUsLinkComponent
Beta Defensin-4 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 50 amino acids and h aving a molecular mass of kDa. The BD-4 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80004-5
Supplier:  Biorbyt
Beta Defensin-4 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 50 amino acids and h aving a molecular mass of kDa. The BD-4 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80004-5
Supplier:  Biorbyt