Order Entry
Italy
ContactUsLinkComponent
TECK Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 127 amino acids and having a m olecular mass of 14.2kDa. The TECK is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80165-20
Supplier:  Biorbyt
TECK Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 127 amino acids and having a m olecular mass of 14.2kDa. The TECK is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80165-20
Supplier:  Biorbyt