Order Entry
Italy
ContactUsLinkComponent
CCL22 Mouse Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 68 amino acids and having a molecular m ass of 7.8kDa. The Mouse CCL22 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80146-5
Supplier:  Biorbyt
CCL22 Mouse Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 68 amino acids and having a molecular m ass of 7.8kDa. The Mouse CCL22 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80146-5
Supplier:  Biorbyt