Order Entry
Italy
ContactUsLinkComponent
MDC Human Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 69 amino acids and having a molecular mas s of 8.1 kDa. The MDC is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80145-5
Supplier:  Biorbyt
MDC Human Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 69 amino acids and having a molecular mas s of 8.1 kDa. The MDC is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80145-5
Supplier:  Biorbyt