Order Entry
Italy
ContactUsLinkComponent
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza virus, strain A/Panama/2007/99. The Influenza Virus was p urified by Ultracentrifugation with 10-40 sucrose gradient. 1 * 50 µG
Catalog # BIRBORB81801-50
Supplier:  Biorbyt
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza virus, strain A/Panama/2007/99. The Influenza Virus was p urified by Ultracentrifugation with 10-40 sucrose gradient. 1 * 50 µG
Catalog # BIRBORB81801-50
Supplier:  Biorbyt