Order Entry
Italy
ContactUsLinkComponent
PSMA Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 150 amino acids (1-150 a.a.) f used to GST tag and having total molecular mass of 43kDa. The PSMA is purified by proprietary chromatographi 1 * 10 µG
Catalog # BIRBORB81287-10
Supplier:  Biorbyt
PSMA Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 150 amino acids (1-150 a.a.) f used to GST tag and having total molecular mass of 43kDa. The PSMA is purified by proprietary chromatographi 1 * 10 µG
Catalog # BIRBORB81287-10
Supplier:  Biorbyt