Order Entry
Italy
ContactUsLinkComponent
Progesterone Receptor Human Recombinant (a.a. 412-562) expressed in E.coli, shows 43 kDa SDS-PAGE (including GST tag).The PGR is pur ified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81275-10
Supplier:  Biorbyt
Progesterone Receptor Human Recombinant (a.a. 412-562) expressed in E.coli, shows 43 kDa SDS-PAGE (including GST tag).The PGR is pur ified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81275-10
Supplier:  Biorbyt