Order Entry
Italy
ContactUsLinkComponent
PDCD1LG2 Human RECOMB produced in E.coli is a single, non-glycosylated, Polypeptide chain fused to GST tag containing 274 amino acid s (1-274) and having total molecular mass of 56kDa. PDCD1LG2 is purified by proprietary chromatographic tech 1 * 10 µG
Catalog # BIRBORB81262-10
Supplier:  Biorbyt
PDCD1LG2 Human RECOMB produced in E.coli is a single, non-glycosylated, Polypeptide chain fused to GST tag containing 274 amino acid s (1-274) and having total molecular mass of 56kDa. PDCD1LG2 is purified by proprietary chromatographic tech 1 * 10 µG
Catalog # BIRBORB81262-10
Supplier:  Biorbyt