Order Entry
Italy
ContactUsLinkComponent
NCF1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 398 amino acids (1-390 a.a.) an d having a molecular mass of 45.7 kDa. The NCF1 is fused to amino acid His-Tag at C-terminus and purified by 1 * 2 µG
Catalog # BIRBORB81229-2
Supplier:  Biorbyt
NCF1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 398 amino acids (1-390 a.a.) an d having a molecular mass of 45.7 kDa. The NCF1 is fused to amino acid His-Tag at C-terminus and purified by 1 * 2 µG
Catalog # BIRBORB81229-2
Supplier:  Biorbyt