Order Entry
Italy
ContactUsLinkComponent
Myoglobin heme free Human Recombinant produced in E.coli is non-glycosylated polypeptide chain having a molecular mass of 11.67 kDa. The Myoglobin heme free contains N-terminal T7 tag and purified by proprietary chromatographic techniques. 1 * 1 mg
Catalog # BIRBORB81228-1
Supplier:  Biorbyt
Myoglobin heme free Human Recombinant produced in E.coli is non-glycosylated polypeptide chain having a molecular mass of 11.67 kDa. The Myoglobin heme free contains N-terminal T7 tag and purified by proprietary chromatographic techniques. 1 * 1 mg
Catalog # BIRBORB81228-1
Supplier:  Biorbyt