Order Entry
Italy
ContactUsLinkComponent
Human Myoglobin produced in Human Cardiac Tissues having a molecular mass of 17.5kDa. Myoglobin is released from recently injured my ocardial cells within few hours of Infarction. Peak levels are reached more quickly than CK-MB or Troponin c 1 * 1 mg
Catalog # BIRBORB81227-1
Supplier:  Biorbyt
Human Myoglobin produced in Human Cardiac Tissues having a molecular mass of 17.5kDa. Myoglobin is released from recently injured my ocardial cells within few hours of Infarction. Peak levels are reached more quickly than CK-MB or Troponin c 1 * 1 mg
Catalog # BIRBORB81227-1
Supplier:  Biorbyt