Order Entry
Italy
ContactUsLinkComponent
MAPT Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 372 amino acids (1-352 a.a.) an d having a molecular mass of 38.9 kDa. The MAPT is fused to 20 amino acid His-Tag at N-terminus and purified 1 * 10 µG
Catalog # BIRBORB81210-10
Supplier:  Biorbyt
MAPT Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 372 amino acids (1-352 a.a.) an d having a molecular mass of 38.9 kDa. The MAPT is fused to 20 amino acid His-Tag at N-terminus and purified 1 * 10 µG
Catalog # BIRBORB81210-10
Supplier:  Biorbyt