Order Entry
Italy
ContactUsLinkComponent
Recombinant Human Holo Lactoferrin produced in Plant is glycosylated polypeptide chain having an approximate molecular mass of 77-80 kDa.The Human Holo Lactotransferrin is purified by proprietary chromatographic techniques. 1 * 10 mg
Catalog # BIRBORB81203-10
Supplier:  Biorbyt
Recombinant Human Holo Lactoferrin produced in Plant is glycosylated polypeptide chain having an approximate molecular mass of 77-80 kDa.The Human Holo Lactotransferrin is purified by proprietary chromatographic techniques. 1 * 10 mg
Catalog # BIRBORB81203-10
Supplier:  Biorbyt