Order Entry
Italy
ContactUsLinkComponent
Recombinant Lamin produced in E.coli is a single, non-glycosylated polypeptide chain containing 645 amino acids and having a molecul ar mass of 70 kDa. Lamin-A is major component of the nuclear lamina, dynamic meshwork located just under the 1 * 2 µG
  BIRBORB81193-2
 :  Biorbyt
Recombinant Lamin produced in E.coli is a single, non-glycosylated polypeptide chain containing 645 amino acids and having a molecul ar mass of 70 kDa. Lamin-A is major component of the nuclear lamina, dynamic meshwork located just under the 1 * 2 µG
  BIRBORB81193-2
 :  Biorbyt
Frequently Bought Together