Order Entry
Italy
ContactUsLinkComponent
CSTB Human Recombinant fused to His-Tag at N-Terminus produced in E.coli is a single, non-glycosylated polypeptide chain containing 118 amino acids and having a molecular mass of 13 kDa. 1 * 5 µG
Catalog # BIRBORB81053-5
Supplier:  Biorbyt
CSTB Human Recombinant fused to His-Tag at N-Terminus produced in E.coli is a single, non-glycosylated polypeptide chain containing 118 amino acids and having a molecular mass of 13 kDa. 1 * 5 µG
Catalog # BIRBORB81053-5
Supplier:  Biorbyt