Order Entry
Italy
ContactUsLinkComponent
SNPH Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 444 amino acids (1-424) and hav ing a molecular mass of 48.2 kDa.The SNPH is fused to 20 amino acid His-Tag at N-terminus and purified by st 1 * 10 µG
Catalog # BIRBORB81374-10
Supplier:  Biorbyt
SNPH Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 444 amino acids (1-424) and hav ing a molecular mass of 48.2 kDa.The SNPH is fused to 20 amino acid His-Tag at N-terminus and purified by st 1 * 10 µG
Catalog # BIRBORB81374-10
Supplier:  Biorbyt