Order Entry
Italy
ContactUsLinkComponent
CD23 Human Recombinant (aa 48-248) expressed in E.coli, shows 48 kDa band on SDS-PAGE. The CD23 is purified by proprietary chromatog raphic techniques. 1 * 10 µG
  BIRBORB81005-10
 :  Biorbyt
CD23 Human Recombinant (aa 48-248) expressed in E.coli, shows 48 kDa band on SDS-PAGE. The CD23 is purified by proprietary chromatog raphic techniques. 1 * 10 µG
  BIRBORB81005-10
 :  Biorbyt
Frequently Bought Together