Order Entry
Italy
ContactUsLinkComponent
RXRA Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 119 amino acids having a molecu lar mass of 13.6 kDa. 1 * 50 µG
  BIRBORB81331-50
 :  Biorbyt
RXRA Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 119 amino acids having a molecu lar mass of 13.6 kDa. 1 * 50 µG
  BIRBORB81331-50
 :  Biorbyt
Frequently Bought Together