YWHAB Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 246 amino acids (1-246) and h aving a molecular mass of 29 kDa. YWHAB is purified by proprietary chromatographic techniques. 1 * 25 µG
Catalog # BIRBORB80879-25
Supplier: Biorbyt
Frequently Bought Together