Order Entry
Italy
ContactUsLinkComponent
STAT1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 732 amino acids (1-712 a.a.) a nd having a molecular mass of 85.2 kDa. The STAT1 is fused to 20 amino acid His-Tag at N-terminus and purifi 1 * 2 µG
Catalog # BIRBORB80870-2
Supplier:  Biorbyt
STAT1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 732 amino acids (1-712 a.a.) a nd having a molecular mass of 85.2 kDa. The STAT1 is fused to 20 amino acid His-Tag at N-terminus and purifi 1 * 2 µG
Catalog # BIRBORB80870-2
Supplier:  Biorbyt