Order Entry
Italy
ContactUsLinkComponent
PGK1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 437 amino acids (1-417 a.a.) a nd having a molecular mass of 46.8kDa. PGK1 is fused to 20 a.a. His-Tag at N-terminus and purified by propri 1 * 25 µG
Catalog # BIRBORB80842-25
Supplier:  Biorbyt
PGK1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 437 amino acids (1-417 a.a.) a nd having a molecular mass of 46.8kDa. PGK1 is fused to 20 a.a. His-Tag at N-terminus and purified by propri 1 * 25 µG
Catalog # BIRBORB80842-25
Supplier:  Biorbyt