Order Entry
Italy
ContactUsLinkComponent
The PI3Ka catalytic and regulatory subunits are coexpressed in Sf9 insect cells. Phosphoinositide 3-kinase alpha Bovine Recombinant is glycosilated protein having molecular weight as follows: p85a chain 83.5 kDa, p110 chain 124.3 kDa. 1 * 2,5 µG
Catalog # BIRBORB80839-2.5
Supplier:  Biorbyt
The PI3Ka catalytic and regulatory subunits are coexpressed in Sf9 insect cells. Phosphoinositide 3-kinase alpha Bovine Recombinant is glycosilated protein having molecular weight as follows: p85a chain 83.5 kDa, p110 chain 124.3 kDa. 1 * 2,5 µG
Catalog # BIRBORB80839-2.5
Supplier:  Biorbyt