Order Entry
Italy
ContactUsLinkComponent
PAK4 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 628 amino acids (1-591 a.a.) an d having a molecular mass of 68.3 kDa.PAK4 is fused to 37 amino acid His-Tag at N-terminus and is purified b 1 * 5 µG
Catalog # BIRBORB80837-5
Supplier:  Biorbyt
PAK4 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 628 amino acids (1-591 a.a.) an d having a molecular mass of 68.3 kDa.PAK4 is fused to 37 amino acid His-Tag at N-terminus and is purified b 1 * 5 µG
Catalog # BIRBORB80837-5
Supplier:  Biorbyt