Order Entry
Italy
ContactUsLinkComponent
p38/SAPK2 is non-glycosilated polypeptide produced by phosphorylation of the purified p38 alpha with MKK6 having a molecular mass of 42.7 kDa. 1 * 1 µG
Catalog # BIRBORB80836-1
Supplier:  Biorbyt
p38/SAPK2 is non-glycosilated polypeptide produced by phosphorylation of the purified p38 alpha with MKK6 having a molecular mass of 42.7 kDa. 1 * 1 µG
Catalog # BIRBORB80836-1
Supplier:  Biorbyt