Order Entry
Italy
ContactUsLinkComponent
Soluble LYVE1 Human Recombinant fused to C-terminal His-tag (6xHis) produced in baculovirus is monomeric, glycosylated, polypeptide containing 232 amino acids (Met-1 to Gly 232) and having total molecular mass of 25 kDa but as result of gly 1 * 2 µG
Catalog # BIRBORB80831-2
Supplier:  Biorbyt
Soluble LYVE1 Human Recombinant fused to C-terminal His-tag (6xHis) produced in baculovirus is monomeric, glycosylated, polypeptide containing 232 amino acids (Met-1 to Gly 232) and having total molecular mass of 25 kDa but as result of gly 1 * 2 µG
Catalog # BIRBORB80831-2
Supplier:  Biorbyt