Order Entry
Italy
ContactUsLinkComponent
HK1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain fused to His tag at the N-terminal encodin g the sequence of 937 amino acids and having a molecular mass of 104.6 kDa.HXK1 is purified by proprietary c 1 * 2 µG
  BIRBORB80820-2
 :  Biorbyt
HK1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain fused to His tag at the N-terminal encodin g the sequence of 937 amino acids and having a molecular mass of 104.6 kDa.HXK1 is purified by proprietary c 1 * 2 µG
  BIRBORB80820-2
 :  Biorbyt
Frequently Bought Together