Order Entry
Italy
ContactUsLinkComponent
Pro BMP -2 Human Recombinant is the precursor of BMP-2 which has intramolecular cystein bridges is homodimer, non-glycosylated, poly peptide chain containing 377 amino acids and having a molecular mass of 85,476 Dalton. Pro BMP is purified b 1 * 10 µG
Catalog # BIRBORB80275-10
Supplier:  Biorbyt
Pro BMP -2 Human Recombinant is the precursor of BMP-2 which has intramolecular cystein bridges is homodimer, non-glycosylated, poly peptide chain containing 377 amino acids and having a molecular mass of 85,476 Dalton. Pro BMP is purified b 1 * 10 µG
Catalog # BIRBORB80275-10
Supplier:  Biorbyt
Frequently Bought Together