Order Entry
Italy
ContactUsLinkComponent
Inhibin-Alpha Human Recombinant produced in E.coli is non-glycosylated, polypeptide chain containing 264 amino acids comprising of b oth and chains, having a molecular mass of 33.5 kDa.The Inhibin-Alpha is fused with an amino-terminal hexahi 1 * 10 µG
Catalog # BIRBORB80734-10
Supplier:  Biorbyt
Inhibin-Alpha Human Recombinant produced in E.coli is non-glycosylated, polypeptide chain containing 264 amino acids comprising of b oth and chains, having a molecular mass of 33.5 kDa.The Inhibin-Alpha is fused with an amino-terminal hexahi 1 * 10 µG
Catalog # BIRBORB80734-10
Supplier:  Biorbyt