Fractalkine Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 76 amino acids fragment (25-100) having total molecular mass of 17.5kDa and fused with 4.5kDa amino-terminal hexahistidine tag.The 1 * 10 µG
Codice catalogo BIRBORB80658-10
Supplier: Biorbyt
Frequently Bought Together