Order Entry
Italy
ContactUsLinkComponent
Glucagon Like Peptide-1 is a single, non-glycosylated, polypeptide containing 30 amino acids and having a molecular mass of 3298.7 Dalton. The GLP-1 is purified by proprietary chromatographic techniques. 1 * 5 mg
Catalog # BIRBORB80723-5
Supplier:  Biorbyt
Glucagon Like Peptide-1 is a single, non-glycosylated, polypeptide containing 30 amino acids and having a molecular mass of 3298.7 Dalton. The GLP-1 is purified by proprietary chromatographic techniques. 1 * 5 mg
Catalog # BIRBORB80723-5
Supplier:  Biorbyt