Order Entry
Italy
ContactUsLinkComponent
IL16 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 150 amino acids and having a mo lecular mass of 15 kDa. The IL-16 is fused to His-Tag at N-Terminus.The IL-16 is purified by proprietary chr 1 * 25 µG
Catalog # BIRBORB80450-25
Supplier:  Biorbyt
IL16 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 150 amino acids and having a mo lecular mass of 15 kDa. The IL-16 is fused to His-Tag at N-Terminus.The IL-16 is purified by proprietary chr 1 * 25 µG
Catalog # BIRBORB80450-25
Supplier:  Biorbyt