Order Entry
Italy
ContactUsLinkComponent
Growth Hormone Releasing Peptide-6 Synthetic is a single, non-glycosylated polypeptide chain containing amino acids, having a molecu lar mass of 873 Dalton and a molecular formula of C46H56N12O6. 1 * 100 mg
Codice catalogo BIRBORB80721-100
Supplier:  Biorbyt
Growth Hormone Releasing Peptide-6 Synthetic is a single, non-glycosylated polypeptide chain containing amino acids, having a molecu lar mass of 873 Dalton and a molecular formula of C46H56N12O6. 1 * 100 mg
Codice catalogo BIRBORB80721-100
Supplier:  Biorbyt