Order Entry
Italy
ContactUsLinkComponent
Ganirelix acetate is synthetic decapeptide with high antagonistic activity against naturally occurring gonadotropin-releasing hormon e (GnRH). Ganirelix acetate is derived from native GnRH with substitutions of amino acids at positions 1, 2, 1 * 500 µG
Catalog # BIRBORB80715-500
Supplier:  Biorbyt
Ganirelix acetate is synthetic decapeptide with high antagonistic activity against naturally occurring gonadotropin-releasing hormon e (GnRH). Ganirelix acetate is derived from native GnRH with substitutions of amino acids at positions 1, 2, 1 * 500 µG
Catalog # BIRBORB80715-500
Supplier:  Biorbyt